Gene Transcripts

Gene Symbol
Mkln1
Entrez Gene ID
27418
Gene Name
muskelin 1, intracellular mediator containing kelch motifs
Gene Aliases
A130067F06Rik, AU015903, BB154892
Chromosome Location
Chr. 6: 31398828 - 31511795 on Build GRCm38
UniGene ID
Mm.238038
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013791.2 NP_038819.1 2, 3 203
GenBank mRNA AK037967.1 - 2, 3 204
AK077541.1 - 2, 3 199
AK155457.1 - 2, 3 184
AK164822.1 - 2, 3 199
AK165239.1 - 2, 3 203
AK171980.1 - 2, 3 187
BC013703.1 - 2, 3 187 3668925
CD550341.1 - 2, 3 296
U72194.1 - 2, 3 186

View Full Product Details