Gene Transcripts
- Gene Symbol
- Commd8
- Entrez Gene ID
- 27784
- Gene Name
- COMM domain containing 8
- Gene Aliases
- C77770, D5Buc26e
- Chromosome Location
- Chr. 5: 72156576 - 72168183 on Build GRCm38
- UniGene ID
- Mm.44223
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178599.4 |
NP_848714.1
|
4, 5 |
572 |
|
GenBank mRNA |
AK009275.1 |
- |
4, 5 |
561 |
|
AK012653.1 |
- |
4, 5 |
558 |
|
AK013015.1 |
- |
5, 6 |
643 |
|
AK013371.1 |
- |
4, 5 |
544 |
|
AK017777.1 |
- |
5, 6 |
586 |
|
AK077934.1 |
- |
4, 5 |
572 |
|
AK151560.1 |
- |
4, 5 |
555 |
|
AK158078.1 |
- |
5, 6 |
655 |
|
AK170011.1 |
- |
4, 5 |
545 |
|
BC023888.1 |
- |
1, 2 |
244 |
|
BC051517.1 |
- |
2, 3 |
332 |
|
BC098183.1 |
- |
4, 5 |
548 |
5150466 |
View Full Product Details