Gene Transcripts

Gene Symbol
Lrtm1
Entrez Gene ID
319476
Gene Name
leucine-rich repeats and transmembrane domains 1
Gene Aliases
A930016D02Rik
Chromosome Location
Chr. 14: 29018208 - 29033642 on Build GRCm38
UniGene ID
Mm.95780
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176920.4 NP_795894.1 2 962
GenBank mRNA AK044492.1 - 2 916
AK138618.1 - 2 780
BC051461.1 - 1 25
BC099587.1 - 2 760 30289234
BC132594.1 - 2 666 40130871
BC145196.1 - 2 666
BC145990.1 - 2 666 40131293

View Full Product Details