Gene Transcripts

Gene Symbol
Sidt1
Entrez Gene ID
320007
Gene Name
SID1 transmembrane family, member 1
Gene Aliases
AW045928, B830021E24Rik
Chromosome Location
Chr. 16: 44240177 - 44333328 on Build GRCm38
UniGene ID
Mm.242144
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159419.1 NP_001152891.1 12 1457
NM_198034.3 NP_932151.2 12 1457
GenBank mRNA AK046836.1 - 12 1817
AK171920.1 - 12 1610
BC025888.1 - 2 107 5039343
BC079584.1 - 12 1457 5704672

View Full Product Details