Gene Transcripts

Gene Symbol
Catsper4
Entrez Gene ID
329954
Gene Name
cation channel, sperm associated 4
Gene Aliases
4933412L05
Chromosome Location
Chr. 4: 134211968 - 134227434 on Build GRCm38
UniGene ID
Mm.79072
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130030.1 NP_001123502.1 6 717
NM_177866.4 NP_808534.1 8 936
GenBank mRNA AK077145.1 - 8 923
BC108974.2 - 8 916 40047424
BC108975.1 - 8 916 40047425
EU781510.1 - 7 813
EU781511.1 - 6 692

View Full Product Details