Gene Transcripts
- Gene Symbol
- Nek5
- Entrez Gene ID
- 330721
- Gene Name
- NIMA (never in mitosis gene a)-related expressed kinase 5
- Gene Aliases
- 6530411J22, BB049667
- Chromosome Location
- Chr. 8: 22073615 - 22125415 on Build GRCm38
- UniGene ID
- Mm.113940
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_177898.4 |
NP_808566.2
|
19 |
1849 |
|
GenBank mRNA |
AK032672.1 |
- |
19 |
1851 |
|
View Full Product Details