Gene Transcripts
- Gene Symbol
- Cmtm1
- Entrez Gene ID
- 100504164
- Gene Name
- CKLF-like MARVEL transmembrane domain containing 1
- Gene Aliases
- CHLFH1a, CKLFH, CKLFH1, Cklfsf1
- Chromosome Location
- Chr. 8: 104293542 - 104310145 on Build GRCm38
- UniGene ID
- Mm.443302
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY241873.1 |
- |
|
570 |
|
AY241874.1 |
- |
|
1004 |
|
AY369078.2 |
- |
|
76 |
|
View Full Product Details