Gene Transcripts

Gene Symbol
Mettl21c
Entrez Gene ID
433294
Gene Name
methyltransferase like 21C
Gene Aliases
A530098C11Rik
Chromosome Location
Chr. 1: 44009399 - 44042626 on Build GRCm38
UniGene ID
Mm.123621
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013799.1 NP_001013821.1 5 818
GenBank mRNA AK041301.1 - 5 818
BC147328.1 - 5 756 9056394
BC147329.1 - 5 756 9056414

View Full Product Details