Gene Transcripts

Gene Symbol
Lrrc32
Entrez Gene ID
434215
Gene Name
leucine rich repeat containing 32
Gene Aliases
AI426318, D11S833Eh, D7H11S833E, EG434215, Garp
Chromosome Location
Chr. 7: 98489301 - 98501831 on Build GRCm38
UniGene ID
Mm.323057
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113379.1 NP_001106850.1 2 1176

View Full Product Details