Gene Transcripts

Gene Symbol
Tfr2
Entrez Gene ID
50765
Gene Name
transferrin receptor 2
Gene Aliases
Trfr2
Chromosome Location
Chr. 5: 137569822 - 137588081 on Build GRCm38
UniGene ID
Mm.21757
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289507.1 NP_001276436.1 13 1767
NM_001289509.1 NP_001276438.1 15 1786
NM_001289511.1 NP_001276440.1 14 1644
NM_015799.4 NP_056614.3 15 1726
GenBank mRNA AF207741.1 - 13 1601
AF222895.1 - 14 1644
AK004848.2 - 15 1702
AK004965.1 - 15 1757
AK155526.1 - 14 1864
AK165320.1 - 13 1595
BC013654.1 - 13 1767 4196597

View Full Product Details