Gene Transcripts

Gene Symbol
Acpp
Entrez Gene ID
56318
Gene Name
acid phosphatase, prostate
Gene Aliases
5'-NT, A030005E02Rik, FRAP, Lap, PAP, Ppal
Chromosome Location
Chr. 9: 104288240 - 104337722 on Build GRCm38
UniGene ID
Mm.19941
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019807.2 NP_062781.2 3 338
NM_207668.2 NP_997551.1 3 338
GenBank mRNA AF210243.1 - 3 291
AK020693.1 - 3 299
AK029273.1 - 3 324
AK076383.1 - 3 345
AK163330.1 - 3 325
BC139826.1 - 3 329 40142642

View Full Product Details