Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022430.2 | NP_071875.2 | 6 | 757 | |
GenBank mRNA | AB026044.1 | - | 6 | 707 | |
AF237906.1 | - | 6 | 695 | ||
AK008099.1 | - | 6 | 696 | ||
AY039038.1 | - | 6 | 695 | ||
BC042719.1 | - | 6 | 743 |