Gene Transcripts

Gene Symbol
Uba5
Entrez Gene ID
66663
Gene Name
ubiquitin-like modifier activating enzyme 5
Gene Aliases
5730525G14Rik, AW240750, Ube1dc1
Chromosome Location
Chr. 9: 104046588 - 104063176 on Build GRCm38
UniGene ID
Mm.224935
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025692.3 NP_079968.2 8 869
GenBank mRNA AK017787.1 - 8 869
AK077825.1 - 8 850
AK151823.1 - 8 826
AK152029.1 - 8 864
AK168907.1 - 8 863
BC019764.1 - 8 746 3989119

View Full Product Details