Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025959.3 | NP_080235.2 | 4, 5 | 270 | |
GenBank mRNA | AK008996.1 | - | 14 | ||
AK012174.1 | - | 4, 5 | 271 | ||
AK014354.1 | - | 4, 5 | 258 | ||
AK144728.1 | - | 267 | |||
AK166961.1 | - | 4, 5 | 255 | ||
BC037099.1 | - | 4, 5 | 268 | ||
BC043044.1 | - | 249 | |||
BC057997.1 | - | 226 | |||
BC116752.1 | - | 4, 5 | 270 | 40126071 | |
BC119168.1 | - | 4, 5 | 270 | 8733917 |