Gene Transcripts
- Gene Symbol
- Kbtbd4
- Entrez Gene ID
- 67136
- Gene Name
- kelch repeat and BTB (POZ) domain containing 4
- Gene Aliases
- 2510026C23Rik
- Chromosome Location
- Chr. 2: 90904724 - 90910561 on Build GRCm38
- UniGene ID
- Mm.330394
- Species
- Mus musculus
- Gene Symbol
- Ptpmt1
- Entrez Gene ID
- 66461
- Gene Name
- protein tyrosine phosphatase, mitochondrial 1
- Gene Aliases
- 1110001D10Rik, 2810004N20Rik, PLIP
- Chromosome Location
- Chr. 2: 90910713 - 90918050 on Build GRCm38
- UniGene ID
- Mm.23926
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK168416.1 |
- |
|
2292 |
|
View Full Product Details