Gene Transcripts

Gene Symbol
Mthfd2l
Entrez Gene ID
665563
Gene Name
methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like
Gene Aliases
1110019K23Rik, C630010D07Rik
Chromosome Location
Chr. 5: 90931196 - 91021368 on Build GRCm38
UniGene ID
Mm.153030
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026788.1 NP_081064.1 2, 3 433
GenBank mRNA BU610697.1 - 1, 2 114
BY014284.1 - 2, 3 349

View Full Product Details