Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027460.2 | NP_081736.2 | 2, 3 | 439 | |
GenBank mRNA | AK017628.1 | - | 2, 3 | 351 | |
AK157423.1 | - | 2, 3 | 440 | ||
AK187296.1 | - | 1, 2 | 25 | ||
BC011293.1 | - | 2, 3 | 414 | 3989313 | |
BQ950692.1 | - | 2, 3 | 374 | ||
CK795124.1 | - | 2, 3 | 448 |