Gene Transcripts

Gene Symbol
Gfod2
Entrez Gene ID
70575
Gene Name
glucose-fructose oxidoreductase domain containing 2
Gene Aliases
5730466C23Rik
Chromosome Location
Chr. 8: 105716110 - 105758668 on Build GRCm38
UniGene ID
Mm.40954
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027469.4 NP_081745.1 3 1255
GenBank mRNA AK017684.1 - 3 1256
AK141521.1 - 1265
BC013623.1 - 3 1267 3969153

View Full Product Details