Gene Transcripts

Gene Symbol
Spata1
Entrez Gene ID
70951
Gene Name
spermatogenesis associated 1
Gene Aliases
4921536I21Rik, AV266943, SP-2
Chromosome Location
Chr. 3: 146457200 - 146499753 on Build GRCm38
UniGene ID
Mm.383235
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027617.3 NP_081893.1 13, 14 1535
GenBank mRNA BC139279.1 - 12, 13 1448
BC139280.1 - 12, 13 1448 8862302
BC171970.1 - 12, 13 1446

View Full Product Details