Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027629.3 | NP_081905.1 | 2 | 569 | |
GenBank mRNA | AK039189.1 | - | 2 | 553 | |
AK040513.1 | - | 2 | 489 | ||
BB633030.1 | - | 2 | 550 | ||
BC020351.1 | - | 2 | 782 | ||
BC076571.1 | - | 2 | 466 | 30356163 | |
CF531609.1 | - | 2 | 466 |