Gene Transcripts

Gene Symbol
Ccdc57
Entrez Gene ID
71276
Gene Name
coiled-coil domain containing 57
Gene Aliases
4933434G05Rik
Chromosome Location
Chr. 11: 120826529 - 120932902 on Build GRCm38
UniGene ID
Mm.275751
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027745.1 NP_082021.1 19 3102
GenBank mRNA AK017054.1 - 4 569
BC030385.1 - 6 889
BC056480.1 - 19 3102 5689542

View Full Product Details