Gene Transcripts

Gene Symbol
Nupl1
Entrez Gene ID
71844
Gene Name
nucleoporin like 1
Gene Aliases
1700017F11Rik, mKIAA0410
Chromosome Location
Chr. 14: 60205610 - 60251502 on Build GRCm38
UniGene ID
Mm.258051
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170591.1 NP_733479.1 13 1566
GenBank mRNA AK006062.1 - 13 1669
AK029688.1 - 1620
AK078279.1 - 13 1614
AK086412.1 - 13 1634
AK129134.1 - 13 1564
AK133721.1 - 13 1577
AK139895.1 - 13 1697
AK146477.1 - 13 1566
AK200201.1 - 3 294
BC026743.1 - 13 1597 3964978

View Full Product Details