Gene Transcripts

Gene Symbol
Utp14a
Entrez Gene ID
72554
Gene Name
UTP14, U3 small nucleolar ribonucleoprotein, homolog A (yeast)
Gene Aliases
2700066J21Rik, JsdX, NY-CO-16, SDCCAG16, T25628, mKIAA0266
Chromosome Location
Chr. X: 48256934 - 48282449 on Build GRCm38
UniGene ID
Mm.336045
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028276.1 NP_082552.1 11 1143
GenBank mRNA AK134784.1 - 11 1168
AK163230.1 - 1 2158
AK172924.1 - 11 1143
AY316163.1 - 11 1166
BC062165.1 - 11 1192
BC082590.1 - 11 1169 30625504

View Full Product Details