Gene Transcripts

Gene Symbol
Hyal4
Entrez Gene ID
77042
Gene Name
hyaluronoglucosaminidase 4
Gene Aliases
4632428M18Rik, CSHY
Chromosome Location
Chr. 6: 24748367 - 24766519 on Build GRCm38
UniGene ID
Mm.66017
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029848.1 NP_084124.1 3 571
GenBank mRNA AB524072.1 - 2 434
AK014599.1 - 594
BC125402.1 - 3 571 40129917
BC132096.1 - 3 571 40130373

View Full Product Details