Gene Transcripts

Gene Symbol
Map7d2
Entrez Gene ID
78283
Gene Name
MAP7 domain containing 2
Gene Aliases
1600028E09Rik, 2900002G04Rik, 5330432J06Rik, C030036K04Rik, Mtap7d2
Chromosome Location
Chr. X: 159414527 - 159498961 on Build GRCm38
UniGene ID
Mm.160079
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081124.1 NP_001074593.1 13 1925
GenBank mRNA AK030316.1 - 9 1427
AK045412.1 - 6 901

View Full Product Details