Gene Transcripts

Gene Symbol
Tex14
Entrez Gene ID
83560
Gene Name
testis expressed gene 14
Gene Aliases
C85585
Chromosome Location
Chr. 11: 87405050 - 87555823 on Build GRCm38
UniGene ID
Mm.440758
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199293.1 NP_001186222.1 8 912
NM_031386.2 NP_113563.2 8 893

View Full Product Details