Gene Transcripts

Gene Symbol
Trim33
Entrez Gene ID
94093
Gene Name
tripartite motif-containing 33
Gene Aliases
8030451N04Rik, AI413936, Ecto, Tif1g
Chromosome Location
Chr. 3: 103279293 - 103358770 on Build GRCm38
UniGene ID
Mm.195036
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079830.2 NP_001073299.1 11 2613
NM_053170.3 NP_444400.2 11 2613
GenBank mRNA AK129293.1 - 11 1872
AY458590.1 - 11 2619

View Full Product Details