Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_053199.3 | NP_444429.1 | 4 | 644 | |
GenBank mRNA | AF195662.1 | - | 4 | 588 | |
AK036352.1 | - | 5 | 700 | ||
AK038917.1 | - | 4 | 601 | ||
AK053077.1 | - | 4 | 598 | ||
AY059393.1 | - | 4 | 443 | ||
BC029659.1 | - | 1 | 63 | ||
BC079837.1 | - | 5 | 684 | ||
BC115417.2 | - | 4 | 549 | 40093410 | |
BC115418.2 | - | 4 | 549 | 40093417 |