Gene Transcripts

Gene Symbol
Stk17b
Entrez Gene ID
98267
Gene Name
serine/threonine kinase 17b (apoptosis-inducing)
Gene Aliases
3110009A03Rik, AI120141, Drak2
Chromosome Location
Chr. 1: 53755512 - 53785250 on Build GRCm38
UniGene ID
Mm.25559
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133810.3 NP_598571.2 8 1281
GenBank mRNA AK029551.1 - 8 1224
AK035693.1 - 8 1249
AK041139.1 - 4 616
AK076932.1 - 8 1248
AK083332.1 - 8 1250
AK153108.1 - 8 1225
AK165291.1 - 8 1221
AY092028.1 - 8 1234
BC006579.1 - 8 1236 3586767

View Full Product Details