Gene Transcripts

Gene Symbol
Wdr47
Entrez Gene ID
99512
Gene Name
WD repeat domain 47
Gene Aliases
1810073M12Rik, AA552998, mKIAA0893
Chromosome Location
Chr. 3: 108591256 - 108645719 on Build GRCm38
UniGene ID
Mm.285968
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181400.3 NP_852065.2 12 2468
GenBank mRNA AK028113.1 - 6 859
BC040337.1 - 12 2329 3586908

View Full Product Details