Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153795.2 | NP_722490.1 | 6 | 947 | |
GenBank mRNA | AK090332.1 | - | 6 | 844 | |
AK142912.1 | - | 6 | 948 | ||
AK149737.1 | - | 6 | 878 | ||
AK158357.1 | - | 6 | 843 | ||
AK169682.1 | - | 6 | 858 | ||
BC025119.1 | - | 6 | 879 | 4190760 | |
BC032204.1 | - | 6 | 923 | 5342463 | |
BC066803.1 | - | 6 | 908 | 30043017 |