Gene Transcripts

Gene Symbol
Gjc2
Entrez Gene ID
118454
Gene Name
gap junction protein, gamma 2
Gene Aliases
B230382L12Rik, Cx47, Gja12
Chromosome Location
Chr. 11: 59175564 - 59183213 on Build GRCm38
UniGene ID
Mm.40016
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080454.4 NP_536702.3 2 922
NM_175452.4 NP_780661.2 2 940
GenBank mRNA AK046421.1 - 2 941
AK046609.1 - 2 923

View Full Product Details