Gene Transcripts

Gene Symbol
Cxcr2
Entrez Gene ID
12765
Gene Name
chemokine (C-X-C motif) receptor 2
Gene Aliases
CD128, CDw128, Cmkar2, Gpcr16, IL-8Rh, IL-8rb, IL8RA, Il8rb, mIL-8RH
Chromosome Location
Chr. 1: 74153994 - 74161246 on Build GRCm38
UniGene ID
Mm.234466
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009909.3 NP_034039.1 2 682
GenBank mRNA AK158234.1 - 3 688
BC051677.1 - 3 655 30023687
D17630.1 - 2 678
L20337.1 - 1 105

View Full Product Details