Gene Transcripts
- Gene Symbol
- Csf2
- Entrez Gene ID
- 12981
- Gene Name
- colony stimulating factor 2 (granulocyte-macrophage)
- Gene Aliases
- Csfgm, GMCSF, Gm-CSf, MGI-IGM
- Chromosome Location
- Chr. 11: 54247270 - 54249899 on Build GRCm38
- UniGene ID
- Mm.4922
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009969.4 |
NP_034099.2
|
3, 4 |
609 |
|
GenBank mRNA |
AK053196.1 |
- |
3, 4 |
609 |
|
BC116878.1 |
- |
3, 4 |
343 |
40126197 |
BC116880.1 |
- |
3, 4 |
343 |
40126199 |
EU366957.1 |
- |
3, 4 |
320 |
|
M11848.1 |
- |
3, 4 |
278 |
|
X02333.1 |
- |
3, 4 |
493 |
|
X03019.1 |
- |
3, 4 |
352 |
|
X03221.1 |
- |
3, 4 |
351 |
|
X05906.1 |
- |
3, 4 |
251 |
|
View Full Product Details