Gene Transcripts

Gene Symbol
Fgfr4
Entrez Gene ID
14186
Gene Name
fibroblast growth factor receptor 4
Gene Aliases
Fgfr-4
Chromosome Location
Chr. 13: 55152818 - 55168759 on Build GRCm38
UniGene ID
Mm.276715
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008011.2 NP_032037.2 14 2074
GenBank mRNA AK084850.1 - 14 2074
AK085723.1 - 14 2072
BC033313.1 - 14 2261 5132831
X57236.1 - 1 83
X59927.1 - 14 2043

View Full Product Details