Gene Transcripts

Gene Symbol
Myh4
Entrez Gene ID
17884
Gene Name
myosin, heavy polypeptide 4, skeletal muscle
Gene Aliases
AI506973, MHC2B, MM, MYH-2B, Minimsc, Minmus, MyHC-IIb, Myhsf
Chromosome Location
Chr. 11: 67237812 - 67260447 on Build GRCm38
UniGene ID
Mm.297382
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010855.3 NP_034985.2 8 837
GenBank mRNA AK138608.1 - 8 838
CR756940.1 - 2 146

View Full Product Details