Gene Transcripts
- Gene Symbol
- Prkacb
- Entrez Gene ID
- 18749
- Gene Name
- protein kinase, cAMP dependent, catalytic, beta
- Gene Aliases
- Pkacb
- Chromosome Location
- Chr. 3: 146729579 - 146812946 on Build GRCm38
- UniGene ID
- Mm.16766
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164198.1 |
NP_001157670.1
|
2, 3 |
375 |
|
NM_001164199.1 |
NP_001157671.1
|
2, 3 |
248 |
|
NM_001164200.1 |
NP_001157672.1
|
2, 3 |
340 |
|
NM_011100.4 |
NP_035230.1
|
2, 3 |
379 |
|
GenBank mRNA |
AK048319.1 |
- |
2, 3 |
371 |
|
AK139419.1 |
- |
2, 3 |
120 |
|
AK148728.1 |
- |
2, 3 |
189 |
|
AK150122.1 |
- |
2, 3 |
341 |
|
AK150134.1 |
- |
2, 3 |
341 |
|
AK163585.1 |
- |
2, 3 |
153 |
|
AK193506.1 |
- |
2, 3 |
122 |
|
AK204631.1 |
- |
2, 3 |
122 |
|
AK216103.1 |
- |
2, 3 |
174 |
|
BC054533.1 |
- |
2, 3 |
363 |
5704796 |
BY129968.1 |
- |
2, 3 |
249 |
|
CB723114.1 |
- |
2, 3 |
375 |
|
J02626.1 |
- |
2, 3 |
308 |
|
JN189786.1 |
- |
2, 3 |
191 |
|
JN189787.1 |
- |
2, 3 |
140 |
|
JN189788.1 |
- |
2, 3 |
115 |
|
View Full Product Details