Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011520.3 | NP_035650.2 | 3 | 880 | |
GenBank mRNA | AK045898.1 | - | 3 | 666 | |
AK129153.1 | - | 3 | 880 | ||
AK155127.1 | - | 3 | 865 | ||
BC054795.1 | - | 3 | 839 | 6834272 | |
BC062093.1 | - | 3 | 776 | 5705893 | |
U52826.1 | - | 3 | 780 |