Gene Transcripts

Gene Symbol
Kbtbd2
Entrez Gene ID
210973
Gene Name
kelch repeat and BTB (POZ) domain containing 2
Gene Aliases
BC022962, Bklhd1, mKIAA1489
Chromosome Location
Chr. 6: 56777524 - 56798285 on Build GRCm38
UniGene ID
Mm.57295
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145958.2 NP_666070.2 4 1188
GenBank mRNA AK028396.1 - 4 1134
AK129374.1 - 4 1098
BC022962.1 - 1 105 5368323

View Full Product Details