Gene Transcripts

Gene Symbol
Zcchc5
Entrez Gene ID
213436
Gene Name
zinc finger, CCHC domain containing 5
Gene Aliases
D430021I08Rik, Gm375, Mar3, Mart3, ZHC5
Chromosome Location
Chr. X: 106837082 - 106840643 on Build GRCm38
UniGene ID
Mm.85581
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199468.1 NP_955762.1 2 487
GenBank mRNA AK084983.1 - 2 394
BC064821.1 - 2 487 30139133

View Full Product Details