Gene Transcripts

Gene Symbol
Phf1
Entrez Gene ID
21652
Gene Name
PHD finger protein 1
Gene Aliases
AW557215, D17Ertd455e, Pcl1, Phf2, Tctex3
Chromosome Location
Chr. 17: 26933127 - 26937908 on Build GRCm38
UniGene ID
Mm.258953
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB011550.1 - 7, 8 895
AK076365.1 - 5, 6 1811
AK133474.1 - 7, 8 863
AK139088.1 - 7, 8 1101
AK144793.1 - 7, 8 1079
AK155268.1 - 5, 6 1840
BC015077.1 - 7, 8 778
BC145595.1 - 6, 7 869
BC145596.1 - 7, 8 784
U81490.1 - 7, 8 835

View Full Product Details