Gene Transcripts

Gene Symbol
Scrn2
Entrez Gene ID
217140
Gene Name
secernin 2
Gene Aliases
AV001119, D11Moh48, SES2
Chromosome Location
Chr. 11: 97029902 - 97033960 on Build GRCm38
UniGene ID
Mm.46189
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146027.2 NP_666139.1 5 779
GenBank mRNA AK075869.1 - 5 743
BC010784.1 - 5 730
BC021346.1 - 5 748 4163541

View Full Product Details