Gene Transcripts

Gene Symbol
Ppp2r5a
Entrez Gene ID
226849
Gene Name
protein phosphatase 2, regulatory subunit B', alpha
Gene Aliases
PR61alpha
Chromosome Location
Chr. 1: 191351981 - 191397041 on Build GRCm38
UniGene ID
Mm.205569
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144880.4 NP_659129.2 5 1300
GenBank mRNA AK145397.1 - 6 686
BC059026.1 - 5 1300 6810754

View Full Product Details