Gene Transcripts
- Gene Symbol
- Ighv6-6
- Entrez Gene ID
- 238427
- Gene Name
- immunoglobulin heavy variable 6-6
- Gene Aliases
- Gm16695, VH10G1
- Chromosome Location
- Chr. 12: 114434788 - 114435087 on Build GRCm38
- UniGene ID
- Mm.422809
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M59921.1 |
- |
|
20 |
|
M59922.1 |
- |
|
20 |
|
- Gene Symbol
- Igh-VJ558
- Entrez Gene ID
- 16061
- Gene Name
- immunoglobulin heavy chain (J558 family)
- Gene Aliases
- AI893585
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.246497
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC049143.1 |
- |
1 |
78 |
|
View Full Product Details