Gene Transcripts

Gene Symbol
Nelfa
Entrez Gene ID
24116
Gene Name
negative elongation factor complex member A, Whsc2
Gene Aliases
Whsc2, Whsc2h
Chromosome Location
Chr. 5: 33898179 - 33936258 on Build GRCm38
UniGene ID
Mm.332320
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011914.2 NP_036044.1 6 804
GenBank mRNA AF101435.1 - 6 762
AK019191.1 - 6 870
AK078760.1 - 6 804
AK088384.1 - 6 820
AK144173.1 - 818
AK146185.1 - 6 856
AK180606.1 - 2 70
BC038003.1 - 6 805 4912719

View Full Product Details