Gene Transcripts

Gene Symbol
Slc26a11
Entrez Gene ID
268512
Gene Name
solute carrier family 26, member 11
Gene Aliases
F630021I08Rik
Chromosome Location
Chr. 11: 119355532 - 119381076 on Build GRCm38
UniGene ID
Mm.31869
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178743.3 NP_848858.2 12 1330
GenBank mRNA AF345196.1 - 12 1330
AK052202.1 - 7 1417
AK089182.1 - 12 1310
BC038604.1 - 12 1323
BC132493.1 - 7 1198 40130770
BC137897.1 - 7 1198 8860916

View Full Product Details