Gene Transcripts

Gene Symbol
Phf21b
Entrez Gene ID
271305
Gene Name
PHD finger protein 21B
Gene Aliases
A730032D07Rik
Chromosome Location
Chr. 15: 84785376 - 84856132 on Build GRCm38
UniGene ID
Mm.101022
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081166.2 NP_001074635.2 6, 7 1157
GenBank mRNA AK042863.1 - 6, 7 748
AK046530.1 - 6, 7 1082
BC067021.1 - 6, 7 801
BC157925.1 - 6, 7 779 9088098
BC158044.1 - 6, 7 779 9007348
BC172019.1 - 6, 7 779 9054305
BC172145.1 - 6, 7 779 9054815

View Full Product Details