Gene Transcripts

Gene Symbol
Smtnl2
Entrez Gene ID
276829
Gene Name
smoothelin-like 2
Gene Aliases
D130058I21Rik
Chromosome Location
Chr. 11: 72389163 - 72411713 on Build GRCm38
UniGene ID
Mm.46732
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177776.3 NP_808444.1 6 1118
GenBank mRNA AK051585.1 - 6 1118
BC037993.1 - 6 1116 5064159

View Full Product Details