Gene Transcripts

Gene Symbol
Larp7
Entrez Gene ID
28036
Gene Name
La ribonucleoprotein domain family, member 7
Gene Aliases
C330027G06Rik, D3Wsu161e
Chromosome Location
Chr. 3: 127536714 - 127553349 on Build GRCm38
UniGene ID
Mm.291032
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138593.2 NP_613059.2 11 1683
GenBank mRNA BC029178.1 - 3 356
BC132266.1 - 3 354
BC132268.1 - 3 354
BM946683.1 - 5 551

View Full Product Details