Gene Transcripts

Gene Symbol
Cabp5
Entrez Gene ID
29865
Gene Name
calcium binding protein 5
Gene Aliases
Chromosome Location
Chr. 7: 13383295 - 13408880 on Build GRCm38
UniGene ID
Mm.103669
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013877.3 NP_038905.1 2, 3 257
GenBank mRNA AB079605.1 - 1, 2 73
AF169161.1 - 2, 3 218
AK044544.1 - 2, 3 258
AK087416.1 - 2, 3 118
BC018253.1 - 2, 3 280 4504283

View Full Product Details